missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Viruses and Bacteria Poster

Uses illustrations and actual photographs.

Manufacturer:  Fisher Science Education AZ-35-1051

 View more versions of this product

Catalog No. S66668


 

Specifications Description & Specifications

Specifications

Science
27 in.
Closer look at the most prevalent disease causing viruses and bacteria known to man
Viruses and Bacteria Poster
88.9cm
Illustrations and actual photographs
68.6cm
Laminated
35 in.