Please login to your online account to display your discounted pricing

Viruses and Bacteria Poster

Uses illustrations and actual photographs.

$16.20 - $25.00

Specifications

Classification Science
For Use With (Application) Illustrations and actual photographs
Height (English) 27 in.
Height (Metric) 68.6cm
Includes Closer look at the most prevalent disease causing viruses and bacteria known to man
View More Specs
Products
Catalog Number Mfr. No. Material Price Quantity    

S66667

 
fisher science education
AZ-35-1050
Paper Each for $16.20

S66668

 
fisher science education
AZ-35-1051
Laminated Each for $25.00
Description & Specifications

Specifications

Classification Science
For Use With (Application) Illustrations and actual photographs
Height (English) 27 in.
Height (Metric) 68.6cm
Includes Closer look at the most prevalent disease causing viruses and bacteria known to man
Width (English) 35 in.
Width (Metric) 88.9cm

  • A close look at some of the most prevalent disease-causing viruses and bacteria
  • Learn how to identify viruses and bacteria and how they harm humans Dimensions: 35W × 27 in.H (88.9 × 68.6cm)