Please login to your online account to display your discounted pricing

Virus Wall Charts


Beautifully illustrated wall charts introduce the microscopic world of viruses.

Manufacturer: fisher science education  WC55

 View more versions of this product

Catalog No. S67832

  • / Each
 

Description & Specifications

Specifications

Product Type Virus I Structure Chart
For Use With (Application) Highlight and provide details for the structure of viruses
Classification Science