missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Virtuoso™ Vial Identification System

Eliminate errors from manual vial labeling. This advanced labeling-information device ensures accurate sample identification and increases productivity.


Products 8
Catalog Number Mfr. No. Item Description Price Quantity  
Catalog Number Mfr. No. Item Description Price Quantity  
03-379-001
View Documents
Thermo Scientific™
60180VT401AP
SureStop amber vials, 9 mm blue caps and rubber/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-007
View Documents
Thermo Scientific™
60180VT406AP
Crimp top amber vials, 11 mm crimp caps and silicone/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-006
View Documents
Thermo Scientific™
60180VT406P
Crimp top clear vials, 11 mm crimp caps and silicone/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-009
View Documents
Thermo Scientific™
60180VT407AP
Snap top amber vials, 11 mm blue snap caps and rubber/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-008
View Documents
Thermo Scientific™
60180VT407P
Snap top clear vials, 11 mm blue snap caps and rubber/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-011
View Documents
Thermo Scientific™
60180VT408AP
Snap top amber vials, 11 mm blue snap caps and silicone/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-000
View Documents
Thermo Scientific™
60180VT401P
SureStop clear vials, 9 mm blue caps and rubber/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
03-379-003
View Documents
Thermo Scientific™
60180VT402AP
SureStop amber vials, 9 mm blue caps and silicone/PTFE seals N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

The Thermo Scientific™ Virtuoso™ Vial Identification System is an error-free option to manual vial labeling that turns a necessary chore into a value-added component within a laboratory workflow. With advanced labeling capabilities, the Virtuoso system ensures accurate vial identification and reliable data, and increases laboratory productivity. Use the system’s touch screen and network capabilities to transfer text, graphics, barcodes, and logos directly onto your vials.

Facilitates Your Labeling Requirements
  • Fully loaded with standard or fully user-defined templates
Increases Productivity
  • Eliminate error-prone manual vial labeling
  • Label a vial in just 5 seconds
  • Improve labeling efficiencies by downloading sample lists
  • Simplify vial sample identification
Produces High-Quality Labeling
  • Resists common laboratory solvents, blurring, and smudging
Provides an Advanced Labeling and information System in One Device
  • Designed by experienced analytical scientists
SDS
Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.