Please login to your online account to display your discounted pricing

Vinyltrimethylsilane, 97%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  200330250

 View more versions of this product

Catalog No. AC200330250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 100.24
Quantity 25mL
Color Clear colorless
EINECS Number 212-042-9
TSCA TSCA
Density 0.68g/mL
Fieser 09,498; 10,444; 12,566; 13,343
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3895 to 1.3915 (20°C, 589nm)
Synonym Ethenyltrimethylsilane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Chemical Name or Material Vinyltrimethylsilane
Name Note 97%
Assay Percent Range 97%
Molecular Formula C5H12Si
Linear Formula H2C=CHSi(CH3)3
MDL Number MFCD00008606
CAS 754-05-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Vinyltrimethylsilane
Name Note 97%
CAS 754-05-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H12Si
Linear Formula H2C=CHSi(CH3)3
Formula Weight 100.24
MDL Number MFCD00008606
Synonym Ethenyltrimethylsilane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents