Learn More
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0249818
Description
Vinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsVinyltrimethylsilane is used in semiconductor processing. It is involved in the Preparation of silyl-ethers by Rh(I) catalysis. Further, it is used in the preparation of trimethylsilylethyl sulfoxides which in the presence of fluoride ion generate sulfenate nucleophiles.
Solubility
Miscible with tetrahydrofuran, diethyl ether, benzene and dichloromethane.
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and strong alkalis (bases).
Specifications
54°C to 56°C | |
-132°C | |
50 g | |
MFCD00008606 | |
956572 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
ethenyl(trimethyl)silane | |
79102 | |
97% | |
1.391 |
Vinyltrimethylsilane | |
754-05-2 | |
C5H12Si | |
UN1993 | |
−24°C (−11°F) | |
Miscible with tetrahydrofuran,diethyl ether,benzene and dichloromethane. | |
C[Si](C)(C)C=C | |
100.236 | |
100.24 | |
Mild | |
0.69 |
Chemical Identifiers
754-05-2 | |
100.236 | |
GCSJLQSCSDMKTP-UHFFFAOYSA-N | |
79102 | |
C[Si](C)(C)C=C |
C5H12Si | |
MFCD00008606 | |
vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane | |
ethenyl(trimethyl)silane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 212-042-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only