Learn More
Vinyltriethylsilane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1246106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
145°C to 146°C | |
1112-54-5 | |
C8H18Si | |
UN1993 | |
25°C (77°F) | |
HBWGDHDXAMFADB-UHFFFAOYSA-N | |
ethenyl(triethyl)silane | |
559195 | |
97% | |
0.771 |
Vinyltriethylsilane | |
5 g | |
MFCD00041345 | |
1743830 | |
vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane # | |
CC[Si](CC)(CC)C=C | |
142.32 | |
142.32 | |
1.434 |
Chemical Identifiers
1112-54-5 | |
142.32 | |
HBWGDHDXAMFADB-UHFFFAOYSA-N | |
559195 | |
CC[Si](CC)(CC)C=C |
C8H18Si | |
MFCD00041345 | |
vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane # | |
ethenyl(triethyl)silane |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only