Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinyltrichlorosilane, 97%

Chemical Identifiers

Chemical Name or Material Vinyltrichlorosilane
CAS 75-94-5
Molecular Formula C2H3Cl3Si
Linear Formula CH2=CHSiCl3
Formula Weight 161.49
MDL Number MFCD00000482
UN Number UN1305
Beilstein 1743440
Synonym Trichlorovinylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0438722

  SDS
alfa aesar™
L0438722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0438736

  SDS
alfa aesar™
L0438736
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.49
Chemical Name or Material Vinyltrichlorosilane
Synonym Trichlorovinylsilane
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 75-94-5
RTECS Number VV6125000
Assay 97%
TSCA TSCA
Quantity 100g
Molecular Formula C2H3Cl3Si
Melting Point −95°C
MDL Number MFCD00000482
Linear Formula CH2=CHSiCl3
Boiling Point 91° to 93°C
Health Hazard 1 Danger
Recommended Storage 2° to 8°C
Health Hazard 2 GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYLTRICHLOROSILANE
Flash Point 10°C (50°F)
UN Number UN1305
Beilstein 1743440
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H311-H314-H318-H302-H332
Highly flammable liquid and vapor.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

2° to 8°C