Please login to your online account to display your discounted pricing

Vinyloxytrimethylsilane, 97%, ACROS Organics™

$44.40 - $44.40

Chemical Identifiers

Chemical Name or Material Vinyloxytrimethylsilane
Name Note 97%
CAS 6213-94-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H12OSi
Linear Formula CH2CHOSi(CH3)3
Formula Weight 116.24
MDL Number MFCD00054764
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338700010

  SDS
thermo fisher scientific
338700010
Glass bottle 1g Each for $44.40

AC338700050

  SDS
thermo fisher scientific
338700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 116.24
Quantity 1g
Color Clear light yellow
EINECS Number 228-281-7
TSCA TSCA
Density 0.7700g/mL
Chemical Name or Material Vinyloxytrimethylsilane
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Refractive Index 1.3885 to 1.3905 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C5H12OSi
Linear Formula CH2CHOSi(CH3)3
MDL Number MFCD00054764
CAS 6213-94-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.