Please login to your online account to display your discounted pricing

Vinylidene chloride, 99.9%, stabilized, ACROS Organics™

$135.54 - $135.54

Chemical Identifiers

Chemical Name or Material Vinylidene chloride
Name Note 99.9%
CAS 75-35-4
CAS 150-76-5
Assay ≥99.85%
Assay Percent Range 99.90%
Molecular Formula C2H2Cl2
Linear Formula H2C=CCl2
Formula Weight 96.94
MDL Number MFCD00011653
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC172290010

  SDS
thermo fisher scientific
172290010
Glass bottle 1L Each for $135.54

AC172290025

  SDS
thermo fisher scientific
172290025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.85%
Physical Form Liquid
Formula Weight 96.94
Quantity 1L
Color Clear colorless
Beilstein 01, 186
Merck Index 15, 10192
RTECS Number KV9275000
EINECS Number 200-864-0
TSCA TSCA
Density 1.218g/mL
Synonym 1, 1-Dichloroethylene
CAS 150-76-5
Refractive Index 1.4244 to 1.4264 (20°C, 589nm)
Hydrochloric Acid 2ppm max.
Packaging Glass bottle
Infrared Spectrum Authentic
Stabilizer 180 to 220ppm MEHQ
Water 0.02% max.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if inhaled.
Extremely flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00011653
Linear Formula H2C=CCl2
Name Note 99.9%
Molecular Formula C2H2Cl2
Assay Percent Range 99.90%
Chemical Name or Material Vinylidene chloride
CAS 75-35-4
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Harmful if inhaled.
Extremely flammable liquid and vapor.