missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vinylferrocene, Thermo Scientific Chemicals
$202.92 - $680.16
Chemical Identifiers
CAS | 1271-51-8 |
---|---|
Molecular Formula | C12H12Fe |
Molecular Weight (g/mol) | 212.07 |
MDL Number | MFCD00001434 |
InChI Key | VGBAECKRTWHKHC-UHFFFAOYSA-N |
IUPAC Name | Vinylferrocene |
SMILES | [Fe].c1cccc1.C=Cc1cccc1 |
Description
Used in the synthesis of 2-aminomethylferrocenes from vinyl ferrocene and lithium amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1271-51-8 | |
212.07 | |
VGBAECKRTWHKHC-UHFFFAOYSA-N | |
[Fe].c1cccc1.C=Cc1cccc1 |
C12H12Fe | |
MFCD00001434 | |
Vinylferrocene |
Specifications
1271-51-8 | |
80°C to 85°C (0.7 mmHg) | |
C12H12Fe | |
1 g | |
VGBAECKRTWHKHC-UHFFFAOYSA-N | |
Vinylferrocene | |
212.07 | |
Vinylferrocene |
52°C to 55°C | |
62°C (144°F) | |
MFCD00001434 | |
Insoluble in water. | |
[Fe].c1cccc1.C=Cc1cccc1 | |
212.07 | |
Powder |
Safety and Handling
P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H228-H302
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 215-041-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only