missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinylene carbonate, 98%, stabilized, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 140890250

Catalog No. AC140890250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Vinylene carbonate
872-36-6
162°C
Authentic
Glass bottle
1.4200 to 1.423
25g
13,342
vinylene carbonate, vinyl carbonate, carbonic acid, cyclic vinylene ester, wln: t5ovoj, vinylenecarbonate, vinyleny carbonate, carbonic acid vinylene, 1,3-dioxo-2-one, vinylene carbonate vc, acmc-209qj3
VAYTZRYEBVHVLE-UHFFFAOYSA-N
1,3-dioxol-2-one
0.1% max. BHT
86.05
Stabilized
1.3600g/mL
73°C
97.5% min. (GC)
C3H2O3
MFCD00005380
19, V,4, 72
1.36
Solubility in water: 11.5g / 100mL
O=C1OC=CO1
86.05
13385
98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposu

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 212-825-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only