Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinylcyclopentane, 99%

Chemical Identifiers

Chemical Name or Material Vinylcyclopentane
CAS 3742-34-5
Molecular Formula C7H12
Formula Weight 96.17
MDL Number MFCD00001386
UN Number UN3295
Synonym Ethenylcyclopentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2623003

  SDS
alfa aesar™
H2623003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2623006

  SDS
alfa aesar™
H2623006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.17
MDL Number MFCD00001386
Health Hazard 1 Danger
Chemical Name or Material Vinylcyclopentane
UN Number UN3295
Assay 99%
Molecular Formula C7H12
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Flash Point 10°C (50°F)
Synonym Ethenylcyclopentane
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Recommended Storage 2° to 8°C
Boiling Point 97°C
CAS 3742-34-5
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.

2° to 8°C