missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vinylcyclopentane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2623006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vinylcyclopentane | |
0.704 | |
10°C (50°F) | |
1.436 | |
5 g | |
vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss | |
C=CC1CCCC1 | |
96.17 | |
96.17 |
3742-34-5 | |
97°C | |
C7H12 | |
MFCD00001386 | |
UN3295 | |
BEFDCLMNVWHSGT-UHFFFAOYSA-N | |
ethenylcyclopentane | |
77345 | |
99% |
Chemical Identifiers
3742-34-5 | |
96.17 | |
BEFDCLMNVWHSGT-UHFFFAOYSA-N | |
77345 | |
C=CC1CCCC1 |
C7H12 | |
MFCD00001386 | |
vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss | |
ethenylcyclopentane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 223-133-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only