Please login to your online account to display your discounted pricing

Vinylbenzyl trimethylammonium chloride, 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC422565000

 View more versions of this product

Catalog No. AC422565000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder and Chunks
Formula Weight 211.73
Quantity 500g
Color White to yellow
Assay Percent Range 97%
Linear Formula CH2CHC6H4CH2N(CH3)3Cl
Chemical Name or Material Vinylbenzyl trimethylammonium chloride
Molecular Formula C12H18ClN
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00080690
CAS 26616-35-3

Specifications

Chemical Name or Material Vinylbenzyl trimethylammonium chloride
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H18ClN
Linear Formula CH2CHC6H4CH2N(CH3)3Cl
Formula Weight 211.73
MDL Number MFCD00080690