Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinyl trifluoroacetate, 99%, stab. with ca 10ppm 3,5-di-tert-butylcatechol

Chemical Identifiers

Chemical Name or Material Vinyl trifluoroacetate
CAS 433-28-3
Name Note stabilized with ≈10ppm 3,5-di-tert-butylcatechol
Molecular Formula CF3CO2CH=CH2
Formula Weight 140.06g/mol
MDL Number MFCD00003561
UN Number UN2924
Beilstein 1758566
Synonym Trifluoroacetic acid vinyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0969006

 
alfa aesar™
L09690-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0969014

 
alfa aesar™
L09690-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Boiling Point 39° to 40°C
Quantity 5g
Flash Point −31°C (−23°F)
Chemical Name or Material Vinyl trifluoroacetate
Name Note stabilized with ≈10ppm 3,5-di-tert-butylcatechol
CAS 433-28-3
Assay 99%
Molecular Formula CF3CO2CH=CH2
Formula Weight 140.06g/mol
MDL Number MFCD00003561
UN Number UN2924
Beilstein 1758566
Synonym Trifluoroacetic acid vinyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C