missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2mmol/g, 100-200 mesh, ACROS Organics™

Chemical Identifiers

MDL Number MFCD03458981
Chemical Name or Material Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC358280010 SDS Thermo Fisher Scientific
358280010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD03458981
Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2 mmol/g, 100-200 mesh

Specifications

crosslinking: 1% DVB
Glass bottle
Beads
White to off-white
Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
0.4 to 1.2mmol/g
100 to 200 mesh
MFCD03458981
1g