Please login to your online account to display your discounted pricing

Vinyl Sulfone polystyrene resin, 1% DVB, 0.4-1.2mmol/g, 100-200 mesh, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  358280010

 View more versions of this product

Catalog No. AC358280010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 1g
Color White to off-white
Packaging Glass bottle
Additional Information crosslinking: 1% DVB
Chemical Name or Material Vinyl Sulfone polystyrene resin
Mesh Size 100 to 200 mesh
Name Note 1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
MDL Number MFCD03458981
Loading 0.4 to 1.2mmol/g

Specifications

Chemical Name or Material Vinyl Sulfone polystyrene resin
Name Note 1% DVB, 0.4-1.2 mmol/g, 100-200 mesh
MDL Number MFCD03458981