Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinyl propionate, 97%, stab.

Chemical Identifiers

Chemical Name or Material Vinyl propionate
CAS 105-38-4
Name Note stabilized
Molecular Formula C5H8O2
Linear Formula CH3CH2CO2CH=CH2
Formula Weight 100.12
MDL Number MFCD00027004
UN Number UN3272
Beilstein 1741731
Synonym Propionic acid vinyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1293130

  SDS
alfa aesar™
L1293130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.12
Chemical Name or Material Vinyl propionate
Assay 97%
MDL Number MFCD00027004
CAS 105-38-4
Molecular Formula C5H8O2
Linear Formula CH3CH2CO2CH=CH2
Beilstein 1741731
Synonym Propionic acid vinyl ester
Melting Point −80°C
Health Hazard 1 Danger
Flash Point 6°C (42°F)
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
RTECS Number UF8575000
TSCA TSCA
Name Note stabilized
UN Number UN3272
Boiling Point 93° to 94°C
Quantity 250g
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.