missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl Myristate (stabilized with MEHQ), TCI America™

Manufacturer:  TCI America M072325ML

 View more versions of this product

Catalog No. M072325ML



Specifications

Specifications

Vinyl Myristate (stabilized with MEHQ)
Colorless
5809-91-6
MFCD00059221
ZQZUENMXBZVXIZ-UHFFFAOYSA-N
ethenyl tetradecanoate
545633
Clear Liquid at 20°C
148°C
25mL
C16H30O2
Myristic Acid Vinyl Ester
CCCCCCCCCCCCCC(=O)OC=C
254.414
254.41
≥99.0% (GC)

Chemical Identifiers

5809-91-6
254.414
ZQZUENMXBZVXIZ-UHFFFAOYSA-N
545633
CCCCCCCCCCCCCC(=O)OC=C
C16H30O2
MFCD00059221
Myristic Acid Vinyl Ester
ethenyl tetradecanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents