missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl Myristate (stabilized with MEHQ) 99.0+%, TCI America™

$670.05 - $670.05

Chemical Identifiers

CAS 5809-91-6
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.414
MDL Number MFCD00059221
InChI Key ZQZUENMXBZVXIZ-UHFFFAOYSA-N
Synonym Myristic Acid Vinyl Ester
PubChem CID 545633
IUPAC Name ethenyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0723500ML View Documents Tci America
M0723500ML
500mL Each for $670.05
 
Specifications

Chemical Identifiers

5809-91-6
254.414
ZQZUENMXBZVXIZ-UHFFFAOYSA-N
545633
CCCCCCCCCCCCCC(=O)OC=C
C16H30O2
MFCD00059221
Myristic Acid Vinyl Ester
ethenyl tetradecanoate

Specifications

148°C
500mL
C16H30O2
Myristic Acid Vinyl Ester
CCCCCCCCCCCCCC(=O)OC=C
254.414
254.41
≥99.0% (GC)
Colorless
5809-91-6
MFCD00059221
ZQZUENMXBZVXIZ-UHFFFAOYSA-N
ethenyl tetradecanoate
545633
Clear Liquid at 20°C
Vinyl Myristate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

TSCA : No