Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinyl methacrylate, 98%, stab. with 200ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material Vinyl methacrylate
CAS 4245-37-8
Name Note stabilized with 200ppm 4-methoxyphenol
Molecular Formula C6H8O2
Formula Weight 112.13
MDL Number MFCD00059329
UN Number UN3272
Beilstein 1743336
Synonym Methacrylic acid vinyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0958406

  SDS
alfa aesar™
L0958406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0958414

  SDS
alfa aesar™
L0958414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.13
Chemical Name or Material Vinyl methacrylate
CAS 4245-37-8
Molecular Formula C6H8O2
MDL Number MFCD00059329
Flash Point 16°C (60°F)
TSCA TSCA
Health Hazard 1 Danger
UN Number UN3272
Quantity 5g
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Odor Pungent
Boiling Point 111° to 112°C
Assay 98%
Synonym Methacrylic acid vinyl ester
Health Hazard 3 GHS P Statement
P210-P305+P351+P338-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
Beilstein 1743336
Name Note stabilized with 200ppm 4-methoxyphenol
Safety and Handling

Danger

GHS P Statement
P210-P305+P351+P338-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.