missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vinyl methacrylate, 98%, stab. with 200ppm 4-methoxyphenol

Manufacturer:  Alfa Aesar™ L0958414

 View more versions of this product

Catalog No. AAL0958414



Specifications

Specifications

Vinyl methacrylate
Pungent
25g
4245-37-8
MFCD00059329
1743336
FFYWKOUKJFCBAM-UHFFFAOYSA-N
ethenyl 2-methylprop-2-enoate
77921
98%
16°C (60°F)
111°C to 112°C
stabilized with 200ppm 4-methoxyphenol
C6H8O2
UN3272
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, vinyl methacrylate, acmc-1cr11
CC(=C)C(=O)OC=C
112.128
112.13

Chemical Identifiers

4245-37-8
112.128
FFYWKOUKJFCBAM-UHFFFAOYSA-N
77921
CC(=C)C(=O)OC=C
C6H8O2
MFCD00059329
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, vinyl methacrylate, acmc-1cr11
ethenyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P305+P351+P338-P302+P352
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Documents