Learn More
Vinyl methacrylate, 98%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0958406
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Vinyl methacrylate | |
4245-37-8 | |
111°C to 112°C | |
Pungent | |
1.435 | |
5 g | |
1743336 | |
FFYWKOUKJFCBAM-UHFFFAOYSA-N | |
ethenyl 2-methylprop-2-enoate | |
77921 | |
98% |
stabilized with 200ppm 4-methoxyphenol | |
0.94 | |
16°C (60°F) | |
C6H8O2 | |
MFCD00059329 | |
UN3272 | |
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11 | |
CC(=C)C(=O)OC=C | |
112.128 | |
112.13 |
Chemical Identifiers
4245-37-8 | |
112.128 | |
FFYWKOUKJFCBAM-UHFFFAOYSA-N | |
77921 | |
CC(=C)C(=O)OC=C |
C6H8O2 | |
MFCD00059329 | |
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11 | |
ethenyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 224-205-1
TSCA : Yes
Recommended Storage : Ambient temperatures