Learn More
Vinyl iodide, tech. 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1265606
Description
Vinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsVinyl iodide is an important intermediate used in organic synthesis, chemical research and pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
It is sensitive to light. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
Vinyl iodide | |
2.08 | |
C2H3I | |
MFCD00039404 | |
1731578 | |
vinyl iodide, ethene, iodo, iodoethylene, vinyl iodide, tech., 1-iodoethylene, iodoethyl, iodo-ethene, 1-iodoethyl, 1-iodoethylene #, vinyl iodide, tech | |
GHXZPUGJZVBLGC-UHFFFAOYSA-N | |
iodoethene | |
68976 | |
153.95 | |
Technical |
593-66-8 | |
55°C to 56°C | |
1.53 | |
5 g | |
Light sensitive | |
Not miscible or difficult to mix in water. | |
IC=C | |
153.95 | |
CHEBI:51315 | |
85% |
Chemical Identifiers
593-66-8 | |
153.95 | |
GHXZPUGJZVBLGC-UHFFFAOYSA-N | |
68976 | |
iodoethene |
C2H3I | |
MFCD00039404 | |
vinyl iodide, ethene, iodo, iodoethylene, vinyl iodide, tech., 1-iodoethylene, iodoethyl, iodo-ethene, 1-iodoethyl, 1-iodoethylene #, vinyl iodide, tech | |
CHEBI:51315 | |
IC=C |
Safety and Handling
GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
EINECSNumber : 209-801-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only