missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vinyl decanoate, 95%, stab., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2908106
Description
It is employed as a intermediates for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediates for pharmaceutical.
Solubility
Soluble in methanol. Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Vinyl decanoate | |
4704-31-8 | |
119°C to 120°C (10 mmHg) | |
C12H22O2 | |
1.435 | |
5 g | |
Soluble in methanol. Not miscible or difficult to mix with water. | |
CCCCCCCCCC(=O)OC=C | |
198.306 | |
198.3 |
stabilized | |
0.886 | |
104°C (219°F) | |
CH3(CH2)8CO2CH=CH2 | |
MFCD00027349 | |
vinyl decanoate, decanoic acid, vinyl ester, vinyl n-decanoate, unii-ff00i399li, decanoic acid, ethenyl ester, n-capric acid vinyl ester, decanoic acid vinyl, acmc-1aqz4, decanoic acid vinyl ester, capric acid vinyl ester = vinyl decenoate | |
CMDXMIHZUJPRHG-UHFFFAOYSA-N | |
ethenyl decanoate | |
62140 | |
95% |
Chemical Identifiers
4704-31-8 | |
198.306 | |
CMDXMIHZUJPRHG-UHFFFAOYSA-N | |
62140 | |
CCCCCCCCCC(=O)OC=C |
C12H22O2 | |
MFCD00027349 | |
vinyl decanoate, decanoic acid, vinyl ester, vinyl n-decanoate, unii-ff00i399li, decanoic acid, ethenyl ester, n-capric acid vinyl ester, decanoic acid vinyl, acmc-1aqz4, decanoic acid vinyl ester, capric acid vinyl ester = vinyl decenoate | |
ethenyl decanoate |
Safety and Handling
EINECSNumber : 225-185-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only