Vinyl-Coated Lead Rings
Stabilize flasks in baths and on the bench.
Manufacturer:  Fisher Science Education F18308-4000
 View more versions of this product
Catalog No. S90140D
Specifications Description & Specifications
Specifications
Accessory
2.8 in.
1000 to 4000mL Sidearm flasks without disconnecting tubing
900g
No
7.11cm
2 lb.