Please login to your online account to display your discounted pricing

Vinyl chloroformate, 99%, stabilized, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC275480100

 View more versions of this product

Catalog No. AC275480100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 106.51
Quantity 10mL
Color Clear colorless
EINECS Number 225-874-2
Density 1.2500g/mL
Beilstein 03, 28
Fieser 08,530
Stabilizer 50 to 100ppm BHT
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.4090 to 1.4110 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Fatal if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Linear Formula CH2=CHOC(O)Cl
Chemical Name or Material Vinyl chloroformate
CAS 5130-24-5
MDL Number MFCD00000641
Molecular Formula C3H3ClO2
Assay Percent Range 99%
Name Note 99%

Specifications

Chemical Name or Material Vinyl chloroformate
Name Note 99%
CAS 5130-24-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H3ClO2
Linear Formula CH2=CHOC(O)Cl
Formula Weight 106.51
MDL Number MFCD00000641
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.

SDS & Safety Documents