missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vinyl Butyrate (stabilized with MEHQ), TCI America™

$45.83 - $377.38

Chemical Identifiers

CAS 123-20-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00059410
InChI Key MEGHWIAOTJPCHQ-UHFFFAOYSA-N
Synonym Butyric Acid Vinyl Ester
PubChem CID 31247
IUPAC Name ethenyl butanoate
SMILES CCCC(=O)OC=C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B076525ML View Documents TCI America
B076525ML
25mL Each for $45.83
 
B0765500ML View Documents TCI America
B0765500ML
500mL Each for $377.38
 
B0765100ML View Documents TCI America
B0765100ML
100mL Each for $126.81
 
Specifications

Chemical Identifiers

123-20-6
114.144
MEGHWIAOTJPCHQ-UHFFFAOYSA-N
31247
CCCC(=O)OC=C
C6H10O2
MFCD00059410
Butyric Acid Vinyl Ester
ethenyl butanoate

Specifications

116°C
25mL
C6H10O2
2838
MEGHWIAOTJPCHQ-UHFFFAOYSA-N
ethenyl butanoate
31247
Clear Liquid at 20°C
Vinyl Butyrate (stabilized with MEHQ)
Colorless
123-20-6
MFCD00059410
Butyric Acid Vinyl Ester
CCCC(=O)OC=C
114.144
114.14
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3697

RTECSNumber : ET7000000

TSCA : No