Learn More
Vinyl acetate, 99+%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140840250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vinyl acetate | |
-93°C | |
0.9300g/mL | |
-8°C | |
99% min. (GC) | |
C4H6O2 | |
CH3COOCH=CH2 | |
02,I,63 | |
0.93 | |
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer | |
XTXRWKRVRITETP-UHFFFAOYSA-N | |
ethenyl acetate | |
3 to 15ppm (hydroquinone) | |
0.43 mPa.s (20°C) | |
86.09 | |
Liquid |
108-05-4,123-31-9 | |
Colorless | |
72°C to 73°C | |
Authentic | |
Glass bottle | |
1.3940 to 1.396 | |
25 mL | |
01,1271; 12,565 | |
15,10189 | |
Solubility in water: 23g/L (20°C). Other solubilities: miscible with ethanol and ether,soluble in acetone,benzene and chloroform | |
CC(=O)OC=C | |
86.09 | |
7904 | |
CHEBI:46916 | |
99+% |
Chemical Identifiers
108-05-4 | |
86.09 | |
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer | |
CHEBI:46916 | |
CC(=O)OC=C |
C4H6O2 | |
XTXRWKRVRITETP-UHFFFAOYSA-N | |
7904 | |
ethenyl acetate |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel u
GHS Signal Word: Danger
EINECSNumber : 203-545-4
RUO – Research Use Only