missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Vinyl 2-Chlorobenzoate (stabilized with HQ) 98.0+%, TCI America™
Supplier: TCI America C104825G
Specifications
Vinyl 2-Chlorobenzoate (stabilized with HQ) | |
Yellow | |
C9H7ClO2 | |
25 g | |
ZJIHUSWGELHYBJ-UHFFFAOYSA-N | |
ethenyl 2-chlorobenzoate | |
587335 | |
≥98.0% (GC) |
15721-27-4 | |
130°C | |
MFCD00059479 | |
2-Chlorobenzoic Acid Vinyl Ester | |
C=COC(=O)C1=CC=CC=C1Cl | |
182.603 | |
182.60 | |
Liquid |
Chemical Identifiers
15721-27-4 | |
182.603 | |
ZJIHUSWGELHYBJ-UHFFFAOYSA-N | |
587335 | |
C=COC(=O)C1=CC=CC=C1Cl |
C9H7ClO2 | |
MFCD00059479 | |
2-Chlorobenzoic Acid Vinyl Ester | |
ethenyl 2-chlorobenzoate |
Safety and Handling
TSCA : No