Please login to your online account to display your discounted pricing

American Educational Products Vinegar, American Educational Products


Quantity 1L
Packaging Cider

Manufacturer: american educational products  1LITER

 View more versions of this product

Catalog No. S80238

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 60.05
Quantity 1L
Color Colorless
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Packaging Cider
Chemical Name or Material Vinegar
Linear Formula CH3COOH
Shelf Life Indefinite
ChemAlert Storage Symbol Gray
Concentration or Composition (by Analyte or Components) 4-8% Acetic Acid Solution
CAS 64-19-7

Specifications

Chemical Name or Material Vinegar
CAS 64-19-7
Linear Formula CH3COOH
Formula Weight 60.05
Accessories
 

Indefinite