Please login to your online account to display your discounted pricing

Vinegar, American Educational Products

$7.90 - $7.90

Chemical Identifiers

Chemical Name or Material Vinegar
CAS 64-19-7
Linear Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S80238

 
american educational products
1LITER
Cider 1L Each for $7.90
Case of 12 Each for $94.90
Description & Specifications

Specifications

Formula Weight 60.05
Quantity 1L
Color Colorless
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Packaging Cider
Chemical Name or Material Vinegar
Linear Formula CH3COOH
Shelf Life Indefinite
ChemAlert Storage Symbol Gray
Concentration or Composition (by Analyte or Components) 4-8% Acetic Acid Solution
CAS 64-19-7
Safety and Handling
 

Indefinite