Please login to your online account to display your discounted pricing

Vinegar


Quantity 3.8L
Packaging Liquid - White

Manufacturer: fisher scientific  S25623A

 View more versions of this product

Catalog No. S25623A

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 60.05
Quantity 3.8L
Color Clear colorless
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 4 to 8% Acetic Acid Solution
Chemical Name or Material Vinegar
Shelf Life 5 years
Assay Percent Range 5% v/v
CAS 64-19-7
ChemAlert Storage Symbol Gray
Packaging Liquid - White
CAS 7732-18-5

Specifications

Chemical Name or Material Vinegar
CAS 64-19-7
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 5% v/v
Formula Weight 60.05
Accessories
 

5 years

SDS & Safety Documents