missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

vinculin Mouse, Clone: 7F9, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $353.80

Specifications

Antigen vinculin
Clone 7F9
Host Species Murine
Species Reactivity Human, Mouse, Rat
Applications Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Microscopy,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122863 Life Technologies
LS14977780
25μg Each for $146.45
501122864 Life Technologies
LS14977782
100μg Each for $353.80
Specifications

Description

The monoclonal antibody 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis.
Description & Specifications

Specifications

vinculin
Murine
Immunocytochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Microscopy,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
7F9
Human, Mouse, Rat
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer