missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vincristine sulfate, Tocris

Disrupts microtubules

Manufacturer:  R & D Systems RDS125710

 View more versions of this product

Catalog No. 125710


 

Specifications Description & Specifications

Specifications

Vincristine sulfate
>96%
923.04
10mg
2068-78-2
C46H56N4O10·H2SO4
22-Oxovincaleukoblastine

Chemical Identifiers

>96%
923.04
Vincristine sulfate
C46H56N4O10·H2SO4
22-Oxovincaleukoblastine

Safety and Handling

Recommended Storage : Desiccate at +4°C