Please login to your online account to display your discounted pricing

Vincristine sulfate, Tocris

Disrupts microtubules

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material Vincristine sulfate
Assay Percent Range >96%
Molecular Formula C46H56N4O10·H2SO4
Formula Weight 923.04
Synonym 22-Oxovincaleukoblastine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125750

 
r & d systems
RDS125750
50mg Each for $794.96

125710

 
r & d systems
RDS125710
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 923.04
Quantity 50mg
Chemical Name or Material Vincristine sulfate
Molecular Formula C46H56N4O10·H2SO4
Assay Percent Range >96%
Recommended Storage Desiccate at +4°C
Synonym 22-Oxovincaleukoblastine
CAS 2068-78-2
Safety and Handling

Desiccate at +4°C