Learn More
Vincristine sulfate, 97+%, Thermo Scientific Chemicals
An anti-tumor alkaloid used in cancer research
Supplier: Thermo Scientific Chemicals J60907MA
Description
Vincristine sulfate, is used as an anticancer agent, microtubule disrupter, Induces apoptosis in human lymphoma cells. Other applications include as a cell cycle arresting, apoptotic inducing alkaloid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in DMSO at 60mg/ml. Soluble in water at 25mg/ml with warming. Very poorly soluble in ethanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents, heavy metal salts.
Specifications
Vincristine Sulfate | |
Powder | |
>300°C | |
MFCD00084729 | |
3924631 | |
Soluble in DMSO at 60mg/ml; Soluble in water at 25mg/ml with warming; Very poorly soluble in ethanol | |
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O | |
126963557 | |
≥98% | |
10 mg |
Vincristine sulfate | |
2068-78-2 | |
C46H58N4O14S | |
UN2811 | |
14,9986 | |
AQTQHPDCURKLKT-DJOZZVTQSA-N | |
923.044 | |
923.04 | |
White | |
273°C to 281°C |
Chemical Identifiers
2068-78-2 | |
923.044 | |
AQTQHPDCURKLKT-DJOZZVTQSA-N | |
CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O |
C46H58N4O14S | |
MFCD00084729 | |
126963557 |
Safety and Handling
GHS H Statement
H360-H341
May damage fertility or the unborn child.
Suspected of causing genetic defects.
P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301-H341-H361fd
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 218-190-0
RTECSNumber : OH6340000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only