Please login to your online account to display your discounted pricing

Vinblastine sulfate, Tocris


Disrupts microtubules

Quantity 10mg

Manufacturer: r & d systems  RDS125610

 View more versions of this product

Catalog No. 125610

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 909.06
Quantity 10mg
Chemical Name or Material Vinblastine sulfate
Assay Percent Range >95%
Molecular Formula C46H58N4O9·H2SO4
CAS 143-67-9
Synonym Vincaleukoblastine
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Vinblastine sulfate
Assay Percent Range >95%
Molecular Formula C46H58N4O9·H2SO4
Formula Weight 909.06
Synonym Vincaleukoblastine
Accessories

Desiccate at +4°C