Please login to your online account to display your discounted pricing

Vinblastine sulfate, Tocris

Disrupts microtubules

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material Vinblastine sulfate
Assay Percent Range >95%
Molecular Formula C46H58N4O9·H2SO4
Formula Weight 909.06
Synonym Vincaleukoblastine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125610

 
r & d systems
RDS125610
10mg Each for $142.08

125650

 
r & d systems
RDS125650
50mg Each for $598.33
Description & Specifications

Specifications

Formula Weight 909.06
Quantity 10mg
Chemical Name or Material Vinblastine sulfate
Assay Percent Range >95%
Molecular Formula C46H58N4O9·H2SO4
CAS 143-67-9
Synonym Vincaleukoblastine
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C