Please login to your online account to display your discounted pricing

Vimentin Mouse, eFluor 660, Clone: V9, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $442.25

Specifications

Antigen Vimentin
Clone V9
Host Species Murine
Species Reactivity Canine, Chicken, Human, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124578

 
life technologies
LS50989780
25μg Each for $175.45

501124579

 
life technologies
LS50989782
100μg Each for $442.25
Description & Specifications

Specifications

Antigen Vimentin
Clone V9
Host Species Murine
Species Reactivity Canine, Chicken, Human, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The V9 monoclonal antibody recognizes human Vimentin, a 57kDa protein that functions as a structural component of intermediate filaments. Vimentin is expressed in cells derived from the mesenchyme but also in specific populations such as radial glia and immature glial cells, pancreatic precursor cells. It is proposed to be a marker of cardiac differentiation. In neural cells, vimentin expression is gradually replaced by neurofilaments. Reports have also shown surface expression of vimentin on activated macrophages, platelets, as well as apoptotic T cells and neutrophils.

This antibody also recognizes canine (dog), rat and chicken vimentin but does not recognize mouse vimentin.