missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vimentin Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

$241.71 - $628.90

Specifications

Antigen Vimentin Ab-2
Clone V9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS129P0 Thermo Scientific™ Lab Vision™
MS129P0
Purified Ab with BSA and Azide 100μL Each for $241.71
MS129P1 Thermo Scientific™ Lab Vision™
MS129P1
Purified Ab with BSA and Azide 500μL Each for $628.90
MS129R7 Thermo Scientific™ Lab Vision™
MS129R7
Ready-to-use for IHC (UVLP) 7mL Each for $353.49
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)


Description & Specifications

Specifications

Vimentin Ab-2
Immunohistochemistry (Paraffin)
Murine
Cytoskeletal Protein
Purified vimentin from pig eye lens
V9
Monoclonal
IVD
Primary