Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Vimentin Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS129P1ABX

 View more versions of this product

Catalog No. MS129P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Immunogen Purified vimentin from pig eye lens
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Gerbil
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Antigen Vimentin Ab-2
Format Concentrated
Isotype IgG2a kappa
Quantity 100µL
Clone V9
Host Species Mouse
Regulatory Status IVD
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cytoskeletal Protein
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)