Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Villin Ab-1, Mouse Monoclonal Antibody

Villin Ab-1, Mouse Monoclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$236.18 - $1,121.14

Specifications

Antigen Villin Ab-1
Clone CWWB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1499P

 
thermo scientific™ lab vision™
MS1499P
1mL Each for $1,121.14

MS1499P0

 
thermo scientific™ lab vision™
MS1499P0
100µL Each for $236.18

MS1499P1

 
thermo scientific™ lab vision™
MS1499P1
500µL Each for $665.49

MS1499R7

 
thermo scientific™ lab vision™
MS1499R7
7mL Each for $563.90
Description & Specifications

Specifications

Antigen Villin Ab-1
Clone CWWB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen Human Villin protein

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Villin Ab-1, Mouse Monoclonal Antibody.

Villin can cap, nucleate, sever and bundle actin in a Ca and phosphoinositide-regulated manner. It is associated with the microvillar actin core bundle of intestinal and renal brush border implicated in adsorption. Villin is composed of six repeats, each containing 150 residues that together constitute the core domain followed by the carboxyl-terminal headpiece domain of 87 residues. The core domain retains the Ca dependent capping nucleating and severing activity, whereas the headpiece domain contributes towards actin filament bundling and binding F-actin, independently of Ca

Host Species: Mouse

Clone: CWWB1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Human Villin protein

Molecular Weight: 92.5kDa

Positive Control: Small intestine and kidney

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)