Please login to your online account to display your discounted pricing

Vileda Professional™ EvoControl™ 200 Microfiber Cleanroom Wipes

Designed for fine wiping as it removes contaminants in one swipe. Vileda Professional™ EvoControl™ 200 Microfiber Cleanroom Wipes are widely used in pharmaceutical, biotech and microelectronics industries.

$93.88 - $156.64

Specifications

Product Type EvoControl 200 Cleanroom Wipes
For Use With (Application) ISO 5, LAR, Production and Labs
Edges Laser cut
Color White
Material Microfiber
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19200666

 
freudenberg
142854
23 x 23cm (9 x 9 in.) Pack of 150 Each for $93.88
Case of 20 Pack for $1,660.00

19200667

 
freudenberg
142855
30.5 x 30.5cm (12 x 12 in.) Pack of 150 Each for $156.64
Case of 20 Pack for $2,769.60
Description & Specifications

Specifications

Product Type EvoControl 200 Cleanroom Wipes
For Use With (Application) ISO 5, LAR, Production and Labs
Edges Laser cut
Color White
Material Microfiber
Packaging Quantity 150/Pk.; 3000/Cs.

  • Cleans glass and stainless steel without leaving residues or streaks
  • Microfiber material reach into microscopic surface pores to remove contamination thoroughly
  • Maximize particle pick up
  • Bacteria removal up to 99.999%
  • Cut fibers are melted together for sealed edge which prevents particle release
  • Disposable precision cleaning wipe
  • Laser cut sealed edge wipe for areas where moderate contamination control is required