Please login to your online account to display your discounted pricing

Vigabatrin, Tocris


GABA-T inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS080850

 View more versions of this product

Catalog No. 080850

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 129.16
CAS 60643-86-9
Chemical Name or Material Vigabatrin
Assay Percent Range >98%
Molecular Formula C6H11NO2
Synonym 4-Aminohexenoic acid
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material Vigabatrin
Assay Percent Range >98%
Molecular Formula C6H11NO2
Formula Weight 129.16
Synonym 4-Aminohexenoic acid
Accessories

Desiccate at +4°C