Please login to your online account to display your discounted pricing

Vigabatrin, Tocris

GABA-T inhibitor

$126.32 - $530.82

Chemical Identifiers

Chemical Name or Material Vigabatrin
Assay Percent Range >98%
Molecular Formula C6H11NO2
Formula Weight 129.16
Synonym 4-Aminohexenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

080810

 
r & d systems
RDS080810
10mg Each for $126.32

080850

 
r & d systems
RDS080850
50mg Each for $530.82
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 129.16
Assay Percent Range >98%
CAS 60643-86-9
Chemical Name or Material Vigabatrin
Synonym 4-Aminohexenoic acid
Molecular Formula C6H11NO2
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C