Please login to your online account to display your discounted pricing

Victoria Pure Blue BO, 0.03% (w/v) Aqueous Solution, Ricca Chemical

$74.67 - $74.67

Chemical Identifiers

Chemical Name or Material Victoria Pure Blue BO
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 2390-60-5
CAS Min % 0.03
CAS Max % 0.03
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R9108000120

 
ricca chemical company
RIR9108000120A
Poly natural 120mL Each for $74.67

R91080004A

 
ricca chemical company
RIR91080004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear blue
Quantity 120mL
Concentration or Composition Notes 0.03% (w/v)
Solvent or Matrix Aqueous Solution
CAS Min % 98.0
CAS 2390-60-5
CAS Min % 0.03
Chemical Name or Material Victoria Pure Blue BO
CAS 7732-18-5
Packaging Poly natural
CAS Max % 100.0
CAS Max % 0.03