Please login to your online account to display your discounted pricing

Victoria Pure Blue B O Dye, 0.03%, LabChem™

$49.74 - $49.74

Chemical Identifiers

Chemical Name or Material Victoria Pure Blue B O Dye
CAS 7732-18-5
CAS Max % 99.97
CAS 2390-60-5
CAS Max % 0.03
Linear Formula C33H40ClN3
Formula Weight 514.15
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC266707

 
lab chem
LC266707
Poly Bottle 125mL Each for $49.74
Description & Specifications

Specifications

Formula Weight 514.15
Chemical Name or Material Victoria Pure Blue B O Dye
Quantity 125mL
Physical Form Liquid
Assay 0.03%
Linear Formula C33H40ClN3
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.97
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 2390-60-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.03
Recommended Storage Room Temperature
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Density 1g/mL
Color Dark, bluish-purple
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature