Please login to your online account to display your discounted pricing
Wheaton™ Vial File™ with Screw Cap: Clear
Clear sample vials with a variety of screw cap options
$228.97 - $228.97
Specifications
Type Sample Vial
For Use With (Application) Used for storing and transporting vials
Material Closure Phenolic
View More Specs
Products
Catalog Number Mfr. No. Capacity Graduated Height (Metric) Price Quantity    
02-912-592
wheaton industries
WW224894
2mL (0.5 dr.) No 38mm Case of 60 Each for $228.97
Description & Specifications
Specifications
Type Sample Vial
For Use With (Application) Used for storing and transporting vials
Material Closure Phenolic
The Vial File provides a compact, easily accessible means of sample storage. The vials are stored in individual partitions within a clear polystyrene case. Foam inserts provide extra protection for the vials.
  • Alpha numerical indexing card allows for identification of each sample vial
  • Vials are manufactured from borosilicate glass which provides superior chemical resistance
  • Cat. Nos. 02-912-591 and -597 feature open-top black phenolic screw cap for easy access with a syringe
  • The Teflon-faced silicone lined screw cap is ideal for low-temperature applications; provides excellent chemical barrier
  • Vials and caps are autoclavable